Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323930.1 | NP_001310859.1 | 3 | 1103 | ||
NM_001323931.1 | NP_001310860.1 | 4 | 1435 | |||
NM_001323932.1 | NP_001310861.1 | 4 | 1266 | |||
NM_001323933.1 | NP_001310862.1 | 5 | 1211 | |||
NM_001323934.1 | NP_001310863.1 | 3 | 870 | |||
NM_001323935.1 | NP_001310864.1 | 3 | 1338 | |||
NM_053052.3 | NP_444280.2 | 3 | 1338 | |||
NR_136654.1 | 2 | 561 | ||||
NR_136655.1 | 3 | 1338 | ||||
NR_136656.1 | 2 | 796 | ||||
NR_136657.1 | 2 | 796 | ||||
XM_006711734.3 | XP_006711797.1 | 3 | 1144 | |||
XM_017000231.1 | XP_016855720.1 | 5 | 4377 | |||
XM_017000232.1 | XP_016855721.1 | 4 | 4183 | |||
GenBank mRNA | AK054633.1 | - | 3 | 898 | ||
AY090635.2 | - | 3 | 1338 | |||
BC001332.1 | - | 2 | 677 | |||
BC004418.1 | - | 2 | 668 | |||
BC007786.2 | - | 3 | 881 | |||
BC011145.1 | - | 3 | 891 | |||
BC014059.2 | - | 2 | 839 | |||
BC018760.1 | - | 3 | 1177 | 4300396 | ||
BC032775.1 | - | 2 | 432 | |||
BX648570.1 | - | 2 | 362 | |||
FJ236305.1 | - | 2 | 839 | |||
GQ472206.1 | - | 3 | 881 |