Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323930.1 | NP_001310859.1 | 4 | 1412 | ||
NM_001323931.1 | NP_001310860.1 | 5 | 1744 | |||
NM_001323932.1 | NP_001310861.1 | 5 | 1575 | |||
NM_001323933.1 | NP_001310862.1 | 6 | 1520 | |||
NM_001323934.1 | NP_001310863.1 | 4 | 1179 | |||
NM_001323935.1 | NP_001310864.1 | 4 | 1647 | |||
NM_053052.3 | NP_444280.2 | 4 | 1647 | |||
NR_136654.1 | 3 | 870 | ||||
NR_136655.1 | 4 | 1647 | ||||
NR_136656.1 | 3 | 1105 | ||||
NR_136657.1 | 3 | 1105 | ||||
XM_006711734.3 | XP_006711797.1 | 4 | 1453 | |||
XM_017000231.1 | XP_016855720.1 | 6 | 4686 | |||
XM_017000232.1 | XP_016855721.1 | 5 | 4492 | |||
GenBank mRNA | AK054633.1 | - | 4 | 1207 | ||
AY090635.2 | - | 4 | 1647 | |||
BC001332.1 | - | 3 | 986 | |||
BC004418.1 | - | 3 | 977 | |||
BC007786.2 | - | 4 | 1190 | |||
BC011145.1 | - | 4 | 1200 | |||
BC014059.2 | - | 3 | 1148 | |||
BC018760.1 | - | 4 | 1486 | 4300396 | ||
BC025231.1 | - | 2 | 149 | |||
BC032775.1 | - | 3 | 741 | |||
BX648570.1 | - | 3 | 671 | |||
FJ236305.1 | - | 3 | 1148 | |||
GQ472206.1 | - | 4 | 1190 |