Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030649.2 | NP_085152.2 | 18 | 1611 | ||
XM_005244715.2 | XP_005244772.1 | 18 | 1647 | |||
XM_005244717.3 | XP_005244774.1 | 6 | 605 | |||
XM_011540606.2 | XP_011538908.1 | 18 | 1660 | |||
XM_011540607.1 | XP_011538909.1 | 18 | 4084 | |||
XM_011540608.2 | XP_011538910.1 | 17 | 1565 | |||
XM_011540609.2 | XP_011538911.1 | 18 | 1661 | |||
XM_017000233.1 | XP_016855722.1 | 18 | 1800 | |||
GenBank mRNA | AB051503.1 | - | 17 | 3276 | ||
AF411981.1 | - | 16 | 1411 | |||
BC004874.2 | - | 3 | 215 | |||
BC047001.1 | - | 8 | 1063 | |||
BC051194.1 | - | 18 | 1611 | 4819254 |