Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122772.2 | NP_001116244.1 | 10, 11 | 2146 | ||
NM_014770.3 | NP_055585.1 | 10, 11 | 1228 | |||
XM_005268625.3 | XP_005268682.1 | 10, 11 | 2272 | |||
XM_005268626.2 | XP_005268683.1 | 10, 11 | 1268 | |||
XM_017018772.1 | XP_016874261.1 | 10, 11 | 2273 | |||
GenBank mRNA | AF384128.1 | - | 10, 11 | 1224 | ||
AF413077.1 | - | 10, 11 | 1138 | |||
AK122827.1 | - | 10, 11 | 1232 | |||
AK292672.1 | - | 10, 11 | 1200 | |||
AY128689.1 | - | 10, 11 | 2146 | |||
BC028020.1 | - | 10, 11 | 1195 | 5218187 | ||
D79989.1 | - | 10, 11 | 1226 |