Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053055.4 | NP_444283.2 | 4 | 975 | ||
XM_011509142.2 | XP_011507444.1 | 5 | 633 | |||
GenBank mRNA | AJ313515.1 | - | 4 | 594 | ||
AK024219.1 | - | 1 | 51 | |||
AK096211.1 | - | 3 | 1720 | |||
AK314852.1 | - | 4 | 548 | |||
BC065277.1 | - | 4 | 634 | 6163471 |