Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053055.4 | NP_444283.2 | 5 | 1007 | ||
XM_011509142.2 | XP_011507444.1 | 6 | 665 | |||
GenBank mRNA | AJ313515.1 | - | 5 | 626 | ||
AK024219.1 | - | 2 | 83 | |||
AK096211.1 | - | 4 | 1752 | |||
AK314852.1 | - | 5 | 580 | |||
BC065277.1 | - | 5 | 666 | 6163471 |