Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166293.1 | NP_001159765.1 | 13 | 1798 | ||
NM_001166294.1 | NP_001159766.1 | 12 | 1666 | |||
NM_001166295.1 | NP_001159767.1 | 12 | 1794 | |||
NM_001166417.1 | NP_001159889.1 | 14 | 1976 | |||
NM_014021.3 | NP_054740.3 | 14 | 1848 | |||
XM_005270427.1 | XP_005270484.1 | 13 | 1737 | |||
XM_005270428.1 | XP_005270485.1 | 13 | 1695 | |||
XM_005270429.1 | XP_005270486.1 | 13 | 1734 | |||
XM_006710331.1 | XP_006710394.1 | 13 | 1655 | |||
XM_011540610.1 | XP_011538912.1 | 14 | 1745 | |||
XM_017000234.1 | XP_016855723.1 | 13 | 1726 | |||
XM_017000235.1 | XP_016855724.1 | 13 | 1701 | |||
XM_017000236.1 | XP_016855725.1 | 13 | 1717 | |||
XM_017000237.1 | XP_016855726.1 | 12 | 1506 | |||
GenBank mRNA | AB023140.1 | - | 13 | 1786 | ||
AK001710.1 | - | 9 | 1008 | |||
AK093173.1 | - | 15 | 1857 | |||
AK292475.1 | - | 13 | 1686 | |||
AK294057.1 | - | 12 | 1553 | |||
AK307336.1 | - | 3 | 269 | |||
AK316472.1 | - | 12 | 1572 | |||
AL133046.1 | - | 14 | 1788 | |||
AY367055.1 | - | 14 | 1711 | |||
AY651262.1 | - | 11 | 1368 | |||
BC033637.1 | - | 13 | 1673 | 5578582 | ||
BC035580.1 | - | 12 | 1607 | |||
BC064389.1 | - | 13 | 1759 | 5745142 |