Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017647.3 | NP_060117.3 | 15 | 2133 | ||
GenBank mRNA | AF327355.1 | - | 14 | 1608 | ||
AK000069.1 | - | 7 | 754 | |||
AK095861.1 | - | 15 | 2133 | |||
AK296715.1 | - | 14 | 1339 | |||
AK307398.1 | - | 14 | 1622 | |||
AK315010.1 | - | 15 | 1558 | |||
AL834482.1 | - | 9 | 1021 | |||
BC000131.1 | - | 10 | 1148 | |||
BC036710.2 | - | 14 | 1517 | 4641078 |