Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319051.1 | NP_001305980.1 | 5 | 1955 | ||
NM_054110.4 | NP_473451.3 | 5 | 1600 | |||
XM_005264852.4 | XP_005264909.1 | 5 | 1132 | |||
XM_017005682.1 | XP_016861171.1 | 5 | 9013 | |||
XM_017005683.1 | XP_016861172.1 | 5 | 1163 | |||
XM_017005684.1 | XP_016861173.1 | 5 | 1163 | |||
GenBank mRNA | AB078149.1 | - | 5 | 1125 | ||
AK094780.1 | - | 8 | 1173 | |||
AK312425.1 | - | 5 | 1601 | |||
AL831925.1 | - | 5 | 1628 | |||
AL832575.2 | - | 5 | 1102 | |||
AL832736.1 | - | 5 | 1570 | |||
AY035399.1 | - | 5 | 1408 | |||
AY358443.1 | - | 5 | 1586 | |||
BC014789.1 | - | 5 | 1550 | 4212702 |