Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PARD3B SIRNA Assays ›
Gene Symbol:
PARD3B
Gene Name:
par-3 family cell polarity regulator beta
Gene Aliases:
ALS2CR19, PAR3B, PAR3L, PAR3beta
Chromosome Location:
Chr.2: 204545793 - 205620162 on Build GRCh38
UniGene ID:
Hs.657382
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42178
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARD3B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001302769.1
NP_001289698.1
13, 14
2140
NM_057177.6
NP_476518.4
13, 14
2140
NM_152526.5
NP_689739.4
12, 13
1954
NM_205863.3
NP_995585.2
13, 14
2140
XM_011510552.2
XP_011508854.1
16, 17
2483
XM_011510553.2
XP_011508855.1
15, 16
7206
XM_017003283.1
XP_016858772.1
12, 13
2430
XM_017003284.1
XP_016858773.1
13, 14
5476
XM_017003285.1
XP_016858774.1
14, 15
7027
XM_017003286.1
XP_016858775.1
12, 13
5372
XM_017003287.1
XP_016858776.1
15, 16
7209
XM_017003288.1
XP_016858777.1
15, 16
7211
XM_017003289.1
XP_016858778.1
10, 11
1587
XM_017003290.1
XP_016858779.1
14, 15
2087
XM_017003291.1
XP_016858780.1
13, 14
1959
XM_017003292.1
XP_016858781.1
13, 14
2021
XM_017003293.1
XP_016858782.1
10, 11
1525
XM_017003294.1
XP_016858783.1
15, 16
7203
GenBank mRNA
AB073472.1
-
15, 16
1933
AB092439.1
-
15, 16
1933
AF428250.1
-
13, 14
1933
AF428251.1
-
12, 13
1747
AF466152.1
-
13, 14
1933
AK056157.1
-
15, 16
2021
AK057965.1
-
8, 9
1298
Back To Top
More Information
Gene Ontology Categories:
Function(s)
cell cycle
cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0