Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288821.1 | NP_001275750.1 | 6 | 901 | ||
NM_001288823.1 | NP_001275752.1 | 7 | 1213 | |||
NM_058170.3 | NP_477518.2 | 6 | 1152 | |||
NR_110210.1 | 8 | 1950 | ||||
NR_110211.1 | 7 | 1889 | ||||
NR_110212.1 | 7 | 1638 | ||||
XM_017000240.1 | XP_016855729.1 | 7 | 1056 | |||
GenBank mRNA | AF397392.2 | - | 6 | 794 | ||
AF397393.2 | - | 7 | 1531 | |||
AF397394.1 | - | 6 | 823 | |||
AF397395.1 | - | 7 | 1560 | |||
AF397396.1 | - | 7 | 884 | |||
AF397397.1 | - | 8 | 1621 | |||
AK095551.1 | - | 4 | 378 | |||
AK095724.1 | - | 6 | 948 | |||
AK300870.1 | - | 6 | 901 | |||
AY358722.1 | - | 6 | 865 | |||
BC022531.1 | - | 6 | 910 | 4812035 |