Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANTXR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145794.1 | NP_001139266.1 | 1 | 890 | ||
NM_058172.5 | NP_477520.2 | 1 | 890 | |||
GenBank mRNA | AB209913.1 | - | 1 | 478 | ||
AY040326.1 | - | 1 | 267 | |||
AY233452.1 | - | 1 | 130 |