Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145806.3 | NP_665805.2 | 3 | 378 | ||
NR_130127.1 | 3 | 378 | ||||
GenBank mRNA | AK091711.1 | - | 3 | 326 | ||
AK292380.1 | - | 3 | 324 | |||
AL530248.3 | - | 3 | 373 | |||
BC015139.1 | - | 3 | 368 | |||
BC019897.1 | - | 3 | 348 | 5123355 | ||
HY159562.1 | - | 3 | 354 | |||
HY179580.1 | - | 3 | 377 |
Gene Symbol: ZNF511-PRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU937518.1 | - | 2 | 150 |