Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024593.1 | NP_001019764.1 | 2 | 266 | ||
GenBank mRNA | AK096884.1 | - | 2 | 267 | ||
AK309870.1 | - | 3 | 322 | |||
BC094693.1 | - | 2 | 245 | 30529259 | ||
BC109255.1 | - | 1 | 179 | |||
BC109256.1 | - | 1 | 179 | |||
BX537678.1 | - | 2 | 317 | |||
CR936824.1 | - | 2 | 248 |