Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSS51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024593.1 | NP_001019764.1 | 6 | 1216 | ||
GenBank mRNA | AK096884.1 | - | 6 | 786 | ||
AK309870.1 | - | 7 | 1272 | |||
BC094693.1 | - | 6 | 1195 | 30529259 | ||
BC109255.1 | - | 5 | 1129 | |||
BC109256.1 | - | 5 | 1129 | |||
BX537678.1 | - | 5 | 2016 | |||
CN395159.1 | - | 1 | 57 | |||
CR936824.1 | - | 6 | 1198 |