Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 4 | 1500 | ||
XM_005269491.1 | XP_005269548.1 | 4 | 1570 | |||
XM_005269492.2 | XP_005269549.1 | 2 | 863 | |||
XM_011539216.1 | XP_011537518.1 | 5 | 2228 | |||
XM_011539217.2 | XP_011537519.1 | 5 | 1923 | |||
XM_011539220.2 | XP_011537522.1 | 5 | 1468 | |||
XM_011539222.2 | XP_011537524.1 | 5 | 1590 | |||
XM_011539223.2 | XP_011537525.1 | 4 | 2278 | |||
XM_011539224.2 | XP_011537526.1 | 4 | 1424 | |||
XM_011539226.1 | XP_011537528.1 | 4 | 1423 | |||
XM_011539227.2 | XP_011537529.1 | 4 | 1756 | |||
XM_011539230.2 | XP_011537532.1 | 3 | 1545 | |||
XM_011539231.1 | XP_011537533.1 | 4 | 3555 | |||
XM_011539232.1 | XP_011537534.1 | 4 | 1222 | |||
XM_011539233.1 | XP_011537535.1 | 3 | 985 | |||
GenBank mRNA | AK057500.1 | - | 4 | 1500 | ||
AK297577.1 | - | 5 | 1700 | |||
BC021140.1 | - | 1 | 14 | |||
BC140898.1 | - | 4 | 1487 | 9021766 | ||
BC146869.1 | - | 4 | 1487 | 9056797 |