Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 7 | 2073 | ||
XM_005269491.1 | XP_005269548.1 | 7 | 2143 | |||
XM_005269492.2 | XP_005269549.1 | 5 | 1436 | |||
XM_011539216.1 | XP_011537518.1 | 8 | 2801 | |||
XM_011539217.2 | XP_011537519.1 | 8 | 2496 | |||
XM_011539220.2 | XP_011537522.1 | 8 | 2041 | |||
XM_011539222.2 | XP_011537524.1 | 8 | 2163 | |||
XM_011539223.2 | XP_011537525.1 | 7 | 2851 | |||
XM_011539224.2 | XP_011537526.1 | 7 | 1997 | |||
XM_011539226.1 | XP_011537528.1 | 7 | 1996 | |||
XM_011539227.2 | XP_011537529.1 | 7 | 2329 | |||
XM_011539230.2 | XP_011537532.1 | 6 | 2118 | |||
XM_011539231.1 | XP_011537533.1 | 7 | 4128 | |||
XM_011539232.1 | XP_011537534.1 | 7 | 1795 | |||
XM_011539233.1 | XP_011537535.1 | 6 | 1558 | |||
XM_017015634.1 | XP_016871123.1 | 7 | 2510 | |||
GenBank mRNA | AF435960.1 | - | 4 | 580 | ||
AK297577.1 | - | 8 | 2273 | |||
AK308357.1 | - | 7 | 1735 | |||
BC021140.1 | - | 4 | 587 | |||
BC140898.1 | - | 7 | 2060 | 9021766 | ||
BC146869.1 | - | 7 | 2060 | 9056797 |