Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153336.2 | NP_699167.2 | 5 | 1267 | ||
XM_017015640.1 | XP_016871129.1 | 5 | 893 | |||
GenBank mRNA | AK094271.1 | - | 4 | 1372 | ||
AK127173.1 | - | 4 | 1386 | |||
BC035344.1 | - | 5 | 1267 | 5185692 |