Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136200.1 | NP_001129672.1 | 1 | 163 | ||
NM_144591.3 | NP_653192.2 | 1 | 163 | |||
GenBank mRNA | AK098071.1 | - | 1 | 111 | ||
AK311742.1 | - | 1 | 93 | |||
AL832477.1 | - | 1 | 151 | |||
BC015994.1 | - | 1 | 115 | 4698381 |
Gene Symbol: BORCS7-ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037644.1 | 1 | 163 |