Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191013.1 | NP_001177942.1 | 6 | 1205 | ||
NM_001191014.1 | NP_001177943.1 | 5 | 742 | |||
NM_001191015.1 | NP_001177944.1 | 4 | 640 | |||
NM_183239.1 | NP_899062.1 | 7 | 1307 | |||
XM_006717624.3 | XP_006717687.1 | 6 | 889 | |||
XM_011539270.2 | XP_011537572.1 | 7 | 2370 | |||
XM_011539273.2 | XP_011537575.1 | 5 | 787 | |||
GenBank mRNA | AK094412.1 | - | 2 | 2504 | ||
AK291886.1 | - | 7 | 995 | |||
AK296266.1 | - | 5 | 592 | |||
AK297522.1 | - | 3 | 496 | |||
AY191318.1 | - | 6 | 675 | |||
AY209189.1 | - | 7 | 999 | |||
AY350731.1 | - | 7 | 999 | |||
BC023583.2 | - | 5 | 708 | |||
BC056918.1 | - | 7 | 1307 | 5165084 |