Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276722.1 | NP_001263651.1 | 3 | 562 | ||
NM_001276723.1 | NP_001263652.1 | 3 | 442 | |||
NM_001276724.1 | NP_001263653.1 | 3 | 437 | |||
NM_138787.3 | NP_620142.2 | 3 | 393 | |||
NR_077243.1 | 3 | 562 | ||||
XM_006718138.3 | XP_006718201.1 | 4 | 463 | |||
XM_011519885.2 | XP_011518187.1 | 4 | 728 | |||
GenBank mRNA | AK290833.1 | - | 3 | 386 | ||
AK299642.1 | - | 3 | 369 | |||
AK302634.1 | - | 3 | 434 | |||
BC051031.1 | - | 3 | 343 | 5122890 | ||
BI831382.1 | - | 3 | 356 | |||
CD101968.1 | - | 3 | 425 | |||
CD678640.1 | - | 3 | 348 |
Gene Symbol: LOC729930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077244.1 | 1 | 295 |