Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004947.4 | NP_004938.1 | 11 | 883 | ||
XM_005264914.3 | XP_005264971.1 | 11 | 1155 | |||
XM_005264915.3 | XP_005264972.1 | 11 | 1155 | |||
XM_005264916.3 | XP_005264973.1 | 11 | 933 | |||
XM_005264917.3 | XP_005264974.1 | 11 | 1155 | |||
XM_005264918.3 | XP_005264975.1 | 11 | 932 | |||
XM_006713008.3 | XP_006713071.1 | 11 | 1155 | |||
XM_006713009.3 | XP_006713072.1 | 11 | 1155 | |||
XM_006713010.3 | XP_006713073.1 | 11 | 918 | |||
XM_017005825.1 | XP_016861314.1 | 11 | 934 | |||
XM_017005826.1 | XP_016861315.1 | 11 | 924 | |||
XM_017005827.1 | XP_016861316.1 | 2 | 75 | |||
GenBank mRNA | AB002297.1 | - | 6 | 491 | ||
AY145303.1 | - | 11 | 860 | |||
AY254099.1 | - | 11 | 883 |