Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098526.1 | NP_001091996.1 | 8 | 1133 | ||
NM_001286570.1 | NP_001273499.1 | 7 | 1070 | |||
NM_001286571.1 | NP_001273500.1 | 7 | 1361 | |||
NM_153206.2 | NP_694938.2 | 7 | 1371 | |||
NR_104479.1 | 7 | 1361 | ||||
XM_005271407.3 | XP_005271464.1 | 7 | 1071 | |||
XM_017017213.1 | XP_016872702.1 | 8 | 1058 | |||
XM_017017214.1 | XP_016872703.1 | 7 | 1018 | |||
XM_017017215.1 | XP_016872704.1 | 6 | 946 | |||
GenBank mRNA | AJ515553.2 | - | 8 | 1053 | ||
AK057590.1 | - | 1 | 636 | |||
AK090409.1 | - | 7 | 3848 | |||
AK094399.1 | - | 7 | 1371 | |||
AK298722.1 | - | 7 | 987 | |||
AK300851.1 | - | 7 | 681 | |||
AK301099.1 | - | 8 | 1050 | |||
AY093686.1 | - | 8 | 1050 | |||
AY138965.1 | - | 8 | 1034 | |||
AY358362.1 | - | 8 | 1043 | |||
BC100794.2 | - | 7 | 1060 | 40000649 | ||
BC100797.1 | - | 7 | 1060 | 40000656 |