Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182496.2 | NP_872302.2 | 3, 4 | 366 | ||
XM_006719229.2 | XP_006719292.1 | 3, 4 | 323 | |||
XM_011537883.2 | XP_011536185.1 | 3, 4 | 396 | |||
XM_011537884.2 | XP_011536186.1 | 4, 5 | 509 | |||
XM_011537886.2 | XP_011536188.1 | 4, 5 | 557 | |||
XM_011537887.2 | XP_011536189.1 | 4, 5 | 594 | |||
XM_011537889.1 | XP_011536191.1 | 3, 4 | 364 | |||
GenBank mRNA | AK097408.1 | - | 3, 4 | 366 | ||
BC095479.1 | - | 3, 4 | 314 | 30389334 |