Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152319.3 | NP_689532.1 | 3, 4 | 236 | ||
XM_005268636.3 | XP_005268693.1 | 2, 3 | 118 | |||
XM_011537896.2 | XP_011536198.1 | 5, 6 | 884 | |||
XM_017018796.1 | XP_016874285.1 | 7, 8 | 1074 | |||
GenBank mRNA | AY255793.1 | - | 2, 3 | 141 | ||
AY255794.1 | - | 2, 3 | 141 | |||
BC031670.2 | - | 2, 3 | 162 | 5165130 | ||
BQ429029.1 | - | 3, 4 | 239 |