Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172681.1 | NP_001166152.1 | 2 | 137 | ||
NM_001172682.1 | NP_001166153.1 | 2 | 70 | |||
NM_152320.2 | NP_689533.2 | 3 | 310 | |||
XM_005268637.3 | XP_005268694.1 | 2 | 117 | |||
XM_005268638.3 | XP_005268695.1 | 2 | 330 | |||
XM_005268639.2 | XP_005268696.1 | 2 | 111 | |||
XM_005268640.4 | XP_005268697.1 | 2 | 310 | |||
XM_011537897.1 | XP_011536199.1 | 2 | 310 | |||
XM_011537898.1 | XP_011536200.1 | 2 | 238 | |||
XM_017018797.1 | XP_016874286.1 | 2 | 161 | |||
XM_017018798.1 | XP_016874287.1 | 2 | 61 | |||
XM_017018799.1 | XP_016874288.1 | 1 | 44 | |||
GenBank mRNA | AK055857.1 | - | 2 | 213 | ||
AK298064.1 | - | 2 | 62 | |||
AY842285.1 | - | 2 | 764 | |||
BC018090.1 | - | 2 | 137 | 4791876 | ||
DB445168.1 | - | 2 | 70 |