Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173353.3 | NP_775489.2 | 3 | 412 | ||
GenBank mRNA | AK094614.1 | - | 3 | 281 | ||
AY098914.1 | - | 3 | 412 | |||
BC114442.1 | - | 3 | 367 | 40083878 | ||
BC114499.1 | - | 3 | 349 | 40083876 |