Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173353.3 | NP_775489.2 | 6 | 914 | ||
GenBank mRNA | AY098914.1 | - | 6 | 914 | ||
BC114442.1 | - | 6 | 869 | 40083878 | ||
BC114499.1 | - | 6 | 851 | 40083876 |