Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203436.2 | NP_982260.2 | 1 | 1744 | ||
GenBank mRNA | AY238895.1 | - | 1 | 1744 |