Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286615.1 | NP_001273544.1 | 25 | 3057 | ||
NM_001286616.1 | NP_001273545.1 | 24 | 2625 | |||
NM_178826.3 | NP_849148.2 | 24 | 2952 | |||
XM_011537911.2 | XP_011536213.1 | 27 | 3052 | |||
XM_011537912.2 | XP_011536214.1 | 27 | 3052 | |||
XM_011537913.2 | XP_011536215.1 | 26 | 3007 | |||
XM_011537914.2 | XP_011536216.1 | 24 | 2563 | |||
XM_011537915.2 | XP_011536217.1 | 25 | 2875 | |||
XM_011537916.2 | XP_011536218.1 | 25 | 2716 | |||
XM_017018808.1 | XP_016874297.1 | 24 | 2563 | |||
XM_017018809.1 | XP_016874298.1 | 26 | 3143 | |||
XM_017018810.1 | XP_016874299.1 | 24 | 2805 | |||
XM_017018811.1 | XP_016874300.1 | 23 | 2760 | |||
XM_017018812.1 | XP_016874301.1 | 23 | 4778 | |||
XM_017018813.1 | XP_016874302.1 | 22 | 2628 | |||
XM_017018814.1 | XP_016874303.1 | 25 | 5017 | |||
XM_017018815.1 | XP_016874304.1 | 25 | 5017 | |||
XM_017018816.1 | XP_016874305.1 | 25 | 3100 | |||
XM_017018817.1 | XP_016874306.1 | 25 | 5052 | |||
GenBank mRNA | AK091540.1 | - | 15 | 1573 | ||
AK091591.1 | - | 24 | 2741 | |||
AK092596.1 | - | 12 | 1139 | |||
AK293810.1 | - | 25 | 2695 | |||
AK316105.1 | - | 25 | 2696 | |||
BC109308.2 | - | 24 | 2610 | 40035759 |