Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001655.2 | NP_001001655.1 | 4 | 998 | ||
NM_001145374.1 | NP_001138846.1 | 4 | 1148 | |||
NM_001145375.1 | NP_001138847.1 | 4 | 895 | |||
NM_001205179.1 | NP_001192108.1 | 3 | 846 | |||
NM_001205180.1 | NP_001192109.1 | 3 | 799 | |||
XM_005253835.4 | XP_005253892.1 | 4 | 1164 | |||
XM_005253836.1 | XP_005253893.1 | 3 | 1092 | |||
GenBank mRNA | AB277859.1 | - | 2 | 562 | ||
AY754389.1 | - | 4 | 983 | |||
BC070489.1 | - | 4 | 950 | 6048174 |