Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166058.1 | NP_001159530.1 | 13 | 1092 | ||
NM_130806.3 | NP_570718.1 | 14 | 1164 | |||
XM_017020389.1 | XP_016875878.1 | 14 | 1153 | |||
GenBank mRNA | AF403384.2 | - | 14 | 1199 | ||
AF453828.1 | - | 14 | 1163 | |||
AY899851.1 | - | 13 | 1021 |