Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017923.1 | NP_001017923.1 | 2 | 452 | ||
GenBank mRNA | AF250395.1 | - | 2 | 436 | ||
AK126985.1 | - | 1 | 208 | |||
BC030258.1 | - | 1 | 190 | |||
BC105030.1 | - | 2 | 445 | 8144033 | ||
BC105032.1 | - | 2 | 445 | 8144035 | ||
BM849739.1 | - | 2 | 461 |