Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174891.3 | NP_777551.2 | 2 | 664 | ||
XM_005267317.3 | XP_005267374.1 | 2 | 1142 | |||
XM_005267318.3 | XP_005267375.1 | 2 | 1142 | |||
GenBank mRNA | AF038188.1 | - | 2 | 228 | ||
AK057249.1 | - | 2 | 664 | |||
BC025175.1 | - | 1 | 99 | |||
BC137495.1 | - | 2 | 607 | 9021495 | ||
BC144671.1 | - | 2 | 607 |