Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADSSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320424.1 | NP_001307353.1 | 12 | 1363 | ||
NM_152328.4 | NP_689541.1 | 12 | 1276 | |||
NM_199165.2 | NP_954634.1 | 12 | 1380 | |||
XM_006720026.3 | XP_006720089.1 | 13 | 1275 | |||
XM_011536412.1 | XP_011534714.1 | 13 | 1349 | |||
XM_011536414.2 | XP_011534716.1 | 13 | 1032 | |||
GenBank mRNA | AK054648.1 | - | 1 | 1548 | ||
AK095921.1 | - | 12 | 1272 | |||
AK096124.1 | - | 10 | 2123 | |||
AK096720.1 | - | 12 | 1314 | |||
AY037159.1 | - | 12 | 1261 | |||
BC032039.1 | - | 12 | 1337 | |||
BC047904.1 | - | 12 | 1276 | 5755025 | ||
BX248286.1 | - | 12 | 1233 |