Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001935.3 | NP_001926.2 | 7 | 1039 | ||
XM_005246371.2 | XP_005246428.1 | 7 | 616 | |||
GenBank mRNA | AB451339.1 | - | 6 | 478 | ||
AB451488.1 | - | 6 | 478 | |||
AK314798.1 | - | 7 | 616 | |||
AY429531.1 | - | 5 | 609 | |||
AY429532.1 | - | 7 | 573 | |||
AY429533.1 | - | 6 | 456 | |||
BC013329.2 | - | 6 | 473 | |||
BC065265.1 | - | 7 | 1039 | 6156880 | ||
M74777.1 | - | 7 | 488 | |||
M80536.1 | - | 6 | 478 | |||
X60708.1 | - | 7 | 553 |