Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 4 | 1379 | ||
NM_203467.1 | NP_982292.1 | 3 | 657 | |||
NR_037792.1 | 6 | 1599 | ||||
NR_037793.1 | 5 | 1487 | ||||
GenBank mRNA | AK293156.1 | - | 4 | 1098 | ||
AY052405.1 | - | 4 | 1065 | |||
BC030142.2 | - | 3 | 593 | |||
BC064133.1 | - | 5 | 1232 | |||
BC093697.1 | - | 3 | 359 | |||
BC107888.1 | - | 3 | 406 | |||
BC112241.1 | - | 3 | 359 | |||
BC139921.1 | - | 4 | 1119 | 40148567 | ||
BQ059807.1 | - | 4 | 394 | |||
BX248298.1 | - | 4 | 1122 | |||
BX648029.1 | - | 6 | 1522 |