Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 1, 2 | 514 | ||
NM_203467.1 | NP_982292.1 | 1, 2 | 514 | |||
NR_037792.1 | 1, 2 | 514 | ||||
GenBank mRNA | AK293156.1 | - | 1, 2 | 233 | ||
AY052405.1 | - | 1, 2 | 200 | |||
BC030142.2 | - | 1, 2 | 450 | |||
BC093697.1 | - | 1, 2 | 216 | |||
BC107888.1 | - | 1, 2 | 263 | |||
BC112241.1 | - | 1, 2 | 216 | |||
BC139921.1 | - | 1, 2 | 254 | 40148567 | ||
BI825814.1 | - | 1, 2 | 441 | |||
BI826606.1 | - | 1, 2 | 515 | |||
BI829295.1 | - | 1, 2 | 451 | |||
BQ059807.1 | - | 1, 2 | 184 | |||
BX248298.1 | - | 1, 2 | 257 |