Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206918.2 | NP_996801.2 | 2 | 816 | ||
XM_006720043.3 | XP_006720106.1 | 3 | 831 | |||
GenBank mRNA | AY541700.1 | - | 2 | 736 | ||
BC063598.1 | - | 2 | 763 | 4775939 |