Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166340.1 | NP_001159812.1 | 2 | 916 | ||
NM_001172109.1 | NP_001165580.1 | 3 | 1070 | |||
NM_001172110.1 | NP_001165581.1 | 3 | 912 | |||
NM_001172111.1 | NP_001165582.1 | 3 | 929 | |||
NM_145204.3 | NP_660205.3 | 2 | 805 | |||
XM_005254157.3 | XP_005254214.1 | 2 | 685 | |||
XM_011521215.2 | XP_011519517.1 | 3 | 1203 | |||
XM_011521217.2 | XP_011519519.1 | 2 | 793 | |||
GenBank mRNA | AF308450.1 | - | 2 | 709 | ||
AK097557.1 | - | 2 | 916 | |||
AK292298.1 | - | 2 | 807 | |||
AY008293.1 | - | 2 | 769 | |||
BC031411.1 | - | 2 | 682 | 5195120 |