Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178859.3 | NP_849190.2 | 3 | 466 | ||
XM_005254159.4 | XP_005254216.1 | 3 | 441 | |||
GenBank mRNA | AI793107.1 | - | 3 | 466 | ||
AK129601.1 | - | 3 | 369 | |||
AY194242.1 | - | 2 | 175 | |||
BC103842.1 | - | 2 | 172 | 40002450 | ||
BC103843.1 | - | 2 | 172 | 40002452 | ||
BC103844.1 | - | 2 | 172 | 40002453 |