Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 2 | 786 | ||
XM_005272421.4 | XP_005272478.1 | 2 | 402 | |||
XM_005272422.3 | XP_005272479.1 | 2 | 166 | |||
XM_005272423.4 | XP_005272480.1 | 2 | 405 | |||
XM_017021923.1 | XP_016877412.1 | 2 | 2235 | |||
GenBank mRNA | AB075851.1 | - | 2 | 772 | ||
AK126887.1 | - | 2 | 787 |