Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001385.2 | NP_001376.1 | 6 | 1201 | ||
XM_005250818.3 | XP_005250875.1 | 6 | 1226 | |||
XM_006716518.3 | XP_006716581.1 | 5 | 1067 | |||
XM_011516903.2 | XP_011515205.1 | 6 | 1227 | |||
XM_017013167.1 | XP_016868656.1 | 6 | 1226 | |||
GenBank mRNA | BC034395.1 | - | 6 | 1165 | 5187250 | |
D78011.1 | - | 6 | 1201 |