Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178452.5 | NP_848547.4 | 5 | 822 | ||
XM_006721129.2 | XP_006721192.1 | 5 | 822 | |||
XM_011522853.2 | XP_011521155.1 | 5 | 822 | |||
XM_011522854.2 | XP_011521156.1 | 5 | 822 | |||
XM_011522855.2 | XP_011521157.1 | 5 | 822 | |||
XM_011522857.2 | XP_011521159.1 | 5 | 822 | |||
XM_011522858.2 | XP_011521160.1 | 5 | 822 | |||
XM_017022918.1 | XP_016878407.1 | 5 | 822 | |||
XM_017022919.1 | XP_016878408.1 | 5 | 1130 | |||
GenBank mRNA | AK057238.1 | - | 5 | 774 | ||
AK058059.1 | - | 1 | 79 | |||
AK310961.1 | - | 5 | 773 | |||
AL833327.1 | - | 5 | 793 | |||
AL833328.1 | - | 5 | 793 | |||
AL833336.1 | - | 5 | 793 | |||
BC024009.2 | - | 5 | 765 | 4822973 | ||
BX648657.1 | - | 5 | 795 |