Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031737.2 | NP_001026907.2 | 3, 4 | 369 | ||
XM_006720838.1 | XP_006720901.1 | 2, 3 | 574 | |||
XM_006720843.3 | XP_006720906.1 | 3, 4 | 731 | |||
XM_011522365.1 | XP_011520667.1 | 2, 3 | 574 | |||
XM_011522369.1 | XP_011520671.1 | 3, 4 | 352 | |||
XM_017022930.1 | XP_016878419.1 | 3, 4 | 469 | |||
GenBank mRNA | AK057178.1 | - | 3, 4 | 469 | ||
AK091831.1 | - | 3, 4 | 352 | |||
AK128538.1 | - | 2, 3 | 576 | |||
AK298111.1 | - | 3, 4 | 333 | |||
AK303991.1 | - | 3, 4 | 340 | |||
AY439221.1 | - | 3, 4 | 263 | |||
BC042110.1 | - | 3, 4 | 355 | 5177109 | ||
DC341680.1 | - | 3, 4 | 369 |