Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284511.1 | NP_001271440.1 | 2 | 609 | ||
NM_001284513.1 | NP_001271442.1 | 2 | 232 | |||
NM_001324118.1 | NP_001311047.1 | 2 | 726 | |||
NM_152341.4 | NP_689554.2 | 2 | 726 | |||
GenBank mRNA | AK054564.1 | - | 2 | 310 | ||
AK291373.1 | - | 2 | 428 | |||
AK295348.1 | - | 1 | 243 | |||
AL832527.1 | - | 2 | 339 | |||
AY424282.1 | - | 2 | 296 | |||
BC033703.1 | - | 2 | 384 | 4154019 | ||
BQ935501.1 | - | 2 | 232 |