Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080510.4 | NP_001073979.3 | 5 | 883 | ||
NM_001206983.2 | NP_001193912.1 | 5 | 918 | |||
NM_001206984.2 | NP_001193913.1 | 5 | 584 | |||
NM_001206985.2 | NP_001193914.1 | 4 | 778 | |||
NM_001206986.2 | NP_001193915.1 | 4 | 813 | |||
NM_001206987.2 | NP_001193916.1 | 4 | 957 | |||
NM_001302703.1 | NP_001289632.1 | 6 | 828 | |||
NM_001302704.1 | NP_001289633.1 | 5 | 688 | |||
NM_001302705.1 | NP_001289634.1 | 5 | 567 | |||
NR_038193.2 | 3 | 540 | ||||
XM_006721674.3 | XP_006721737.1 | 5 | 946 | |||
XM_006721675.1 | XP_006721738.1 | 5 | 810 | |||
XM_006721676.3 | XP_006721739.1 | 5 | 845 | |||
XM_006721678.3 | XP_006721741.1 | 5 | 931 | |||
XM_006721679.3 | XP_006721742.1 | 4 | 576 | |||
XM_006721680.2 | XP_006721743.1 | 4 | 485 | |||
XM_011524282.1 | XP_011522584.1 | 5 | 674 | |||
XM_017024145.1 | XP_016879634.1 | 5 | 921 | |||
XM_017024146.1 | XP_016879635.1 | 5 | 688 | |||
GenBank mRNA | BC033998.2 | - | 4 | 754 | ||
BC045819.1 | - | 5 | 845 | |||
BC106939.2 | - | 3 | 406 | |||
BI225157.1 | - | 4 | 778 | |||
BI829268.1 | - | 4 | 741 | |||
BU176249.1 | - | 5 | 570 | |||
BX356072.2 | - | 5 | 667 | |||
BX403283.2 | - | 5 | 694 | |||
CN372746.1 | - | 3 | 496 | |||
JQ313903.1 | - | 4 | 513 |