Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198844.2 | NP_942141.2 | 7 | 1039 | ||
NM_199321.2 | NP_955353.1 | 8 | 1105 | |||
GenBank mRNA | AK292383.1 | - | 8 | 1143 | ||
AY251603.1 | - | 8 | 1088 | |||
AY251604.1 | - | 7 | 1022 | |||
BC043152.2 | - | 7 | 1025 | 5297840 |