Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144607.4 | NP_653208.2 | 4 | 1001 | ||
XM_005256440.3 | XP_005256497.1 | 4 | 712 | |||
GenBank mRNA | AK057061.1 | - | 4 | 933 | ||
AK125598.1 | - | 1 | 595 | |||
AK289520.1 | - | 4 | 684 | |||
BC060779.1 | - | 4 | 753 | 30349502 |