Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152344.3 | NP_689557.1 | 4, 5 | 685 | ||
GenBank mRNA | AK055218.1 | - | 3, 4 | 413 | ||
AL832076.1 | - | 1 | 374 | |||
BC016791.2 | - | 1 | 186 | |||
BC044587.1 | - | 3, 4 | 413 | 4794614 | ||
BC105288.1 | - | 3, 4 | 427 | 6385345 |